Assay Details
| Gene Symbol: Cpox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037095.1 | NP_001032172.1 | 3 - 4 | 857 | 108 | |
| GenBank mRNA | BC107664.1 | - | 3 - 4 | 857 | 7936479 | 108 |
More Information
Updated Mapping Information
Set Membership: |
|

Cpox
Genomic Map