Assay Details
| Gene Symbol: Rpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC058473.1 | - | 1 - 2 | 44 | 6918141 | 74 |
| FQ209984.1 | - | 1 - 2 | 38 | 74 | ||
| FQ220432.1 | - | 1 - 2 | 48 | 74 | ||
| FQ221142.1 | - | 1 - 2 | 37 | 74 | ||
| FQ221221.1 | - | 1 - 2 | 44 | 74 | ||
| FQ221486.1 | - | 1 - 2 | 40 | 74 | ||
| FQ221688.1 | - | 1 - 2 | 40 | 74 | ||
| FQ221810.1 | - | 1 - 2 | 40 | 74 | ||
| FQ221946.1 | - | 1 - 2 | 40 | 74 | ||
| FQ221988.1 | - | 1 - 2 | 44 | 74 | ||
| FQ222823.1 | - | 1 - 2 | 40 | 74 | ||
| FQ223282.1 | - | 1 - 2 | 40 | 74 | ||
| FQ224199.1 | - | 1 - 2 | 40 | 74 | ||
| FQ224575.1 | - | 1 - 2 | 44 | 74 | ||
| FQ224676.1 | - | 1 - 2 | 40 | 74 | ||
| Gene Symbol: RGD1560821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257178.2 | XP_006257240.1 | 1 | 23 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058473.1,XM_006257178.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl24
Genomic Map