Assay Details
| Gene Symbol: Gtf2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100556.1 | NP_001094026.1 | 2 - 3 | 570 | 84 | |
| XM_006248398.2 | XP_006248460.1 | 2 - 3 | 900 | 84 | ||
| XM_008768731.1 | XP_008766953.1 | 2 - 3 | 1422 | 84 | ||
| GenBank mRNA | BC099237.1 | - | 1 - 2 | 109 | 84 | |
| DY311373.1 | - | 2 - 3 | 605 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2e1
Genomic Map