Assay Details
| Gene Symbol: Tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105889.1 | NP_001099359.1 | 2 - 3 | 284 | 106 | |
| XM_008768632.1 | XP_008766854.1 | 2 - 3 | 521 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem30c
Genomic Map