Assay Details
| Gene Symbol: Apod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012777.1 | NP_036909.1 | 2 - 3 | 196 | 76 | |
| GenBank mRNA | FQ212999.1 | - | 2 - 3 | 196 | 76 | |
| X55572.1 | - | 2 - 3 | 196 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apod
Genomic Map