Assay Details
Gene Symbol: Tomm70a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212519.2 | NP_997684.1 | 11 - 12 | 1816 | 95 | |
GenBank mRNA | AB162856.1 | - | 11 - 12 | 1679 | 95 | |
BC098640.1 | - | 11 - 12 | 1816 | 7099109 | 95 | |
FQ228582.1 | - | 1861 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P517682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P125530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10888 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P517682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125530 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P517682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125530 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P517682 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P125530 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P517686 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P517682 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18240 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66663 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10753626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753619 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753617 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753623 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753622 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753621 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753616 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753620 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753628 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753624 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884212 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10753614 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10884211 | - | - |
Rat Genome 230 2.0 Array | 1379186_at | - | - |
Rat Genome 230 2.0 Array | 1373039_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373039_at | - | - |
Rat Genome 230 2.0 Array | 1381258_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379186_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381258_at | - | - |
Affymetrix HT Rat Focus Array | 1379186_at | - | - |
Affymetrix HT Rat Focus Array | 1379186_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100008844.rn.1 | TC1100001462.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100008843.rn.1 | TC1100001462.rn.1 | RTA TC |
Rat Expression Set 230 | 1379186_PM_at | - | - |
Rat Expression Set 230 | 1379186_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753614 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884211 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20914395 | - | - |
ABI Rat Genome Survey Microarray | 21051972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362445 | - | - |
RatRef-12 Expression BeadChip | ILMN_51865 | - | - |
