Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014030.1 | NP_001014052.1 | 1 - 2 | 59 | 61 | |
| XM_006231506.2 | XP_006231568.1 | 1 - 2 | 610 | 61 | ||
| XM_008760434.1 | XP_008758656.1 | 1 - 2 | 611 | 61 | ||
| GenBank mRNA | BC083852.1 | - | 1 - 2 | 59 | 7123646 | 61 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC083852.1,NM_001014030.1,XM_006231506.2,XM_008760434.1 transcripts.
Set Membership: |
|

Tmem180
Genomic Map