Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014030.1 | NP_001014052.1 | 2 - 3 | 281 | 122 | |
| XM_006231506.2 | XP_006231568.1 | 2 - 3 | 832 | 122 | ||
| XM_006231507.2 | XP_006231569.1 | 2 - 3 | 516 | 122 | ||
| XM_008760432.1 | XP_008758654.1 | 4 - 5 | 973 | 122 | ||
| GenBank mRNA | BC083852.1 | - | 2 - 3 | 281 | 7123646 | 122 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem180
Genomic Map