Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231506.2 | XP_006231568.1 | 6 - 7 | 1588 | 63 | |
| XM_006231507.2 | XP_006231569.1 | 6 - 7 | 1272 | 63 | ||
| XM_008760432.1 | XP_008758654.1 | 8 - 9 | 1729 | 63 | ||
| XM_008760433.1 | XP_008758655.1 | 5 - 6 | 1189 | 63 | ||
| XM_008760434.1 | XP_008758656.1 | 5 - 6 | 1506 | 63 | ||
More Information
Set Membership: |
|

Tmem180
Genomic Map