Assay Details
| Gene Symbol: Cyp17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012753.2 | NP_036885.1 | 2 - 3 | 465 | 73 | |
| XM_006231434.2 | XP_006231496.1 | 3 - 4 | 788 | 73 | ||
| XM_006231435.2 | XP_006231497.1 | 3 - 4 | 757 | 73 | ||
| GenBank mRNA | BC078898.1 | - | 2 - 3 | 465 | 7111017 | 73 |
| M22204.1 | - | 2 - 3 | 467 | 73 | ||
| M31681.1 | - | 2 - 3 | 477 | 73 | ||
| X14086.1 | - | 2 - 3 | 467 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp17a1
Genomic Map