Assay Details
Gene Symbol: Spsb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106849.2 | NP_001100319.1 | 1 - 2 | 553 | 81 | |
GenBank mRNA | BC168912.1 | - | 1 - 2 | 553 | 7191741 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC168912.1, NM_001106849.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
