Assay Details
Gene Symbol: Spsb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106849.2 | NP_001100319.1 | 2 - 3 | 1404 | 64 | |
GenBank mRNA | BC168912.1 | - | 2 - 3 | 1404 | 7191741 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P289132 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P289125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80437 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P289125 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P289132 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P289125 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P289132 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P289132 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P289125 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P289132 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P289125 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44991 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24540 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10919430 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919432 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919431 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10919429 | - | - |
Rat Genome 230 2.0 Array | 1384243_at | - | - |
Rat Genome 230 2.0 Array | 1384243_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384243_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384243_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800025438.rn.1 | TC0800003187.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800025439.rn.1 | TC0800003187.rn.1 | RTA TC |
Rat Expression Set 230 | 1384243_PM_at | - | - |
Rat Expression Set 230 | 1384243_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919429 | - | - |
Rat Genome U34 Set | rc_AI111413_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21553073 | - | - |
