Assay Details
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080902.4 | NP_543178.2 | 2 - 3 | 177 | 70 | |
GenBank mRNA | AY062253.3 | - | 2 - 3 | 236 | 70 | |
BC059118.1 | - | 2 - 3 | 181 | 6890532 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
