Assay Details
| Gene Symbol: Tmie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109299.1 | NP_001102769.1 | 4 - 5 | 1525 | 66 | |
| XM_006243995.2 | XP_006244057.1 | 2 - 3 | 1659 | 66 | ||
| XM_006243996.2 | XP_006244058.1 | 2 - 3 | 1301 | 66 | ||
| XM_008766649.1 | XP_008764871.1 | 2 - 3 | 1691 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmie
Genomic Map