Assay Details
| Gene Symbol: Hyal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207599.2 | NP_997482.2 | 2 - 3 | 1030 | 79 | |
| GenBank mRNA | AB100602.1 | - | 1 - 2 | 894 | 79 | |
| BC097259.1 | - | 2 - 3 | 1030 | 7453545 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hyal3
Genomic Map