Assay Details
| Gene Symbol: Cnot10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007003.1 | NP_001007004.1 | 16 - 17 | 2204 | 69 | |
| GenBank mRNA | BC083782.1 | - | 16 - 17 | 2204 | 7096219 | 69 |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P404538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126929 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P404538 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P404538 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P404538 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P404538 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1023173 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22798 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50056 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18102 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10920690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920687 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920688 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920677 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920689 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920680 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10920676 | - | - |
| Rat Genome 230 2.0 Array | 1372988_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372988_at | - | - |
| Affymetrix HT Rat Focus Array | 1372988_at | - | - |
| Affymetrix HT Rat Focus Array | 1372988_PM_at | - | - |
| Rat Expression Set 230 | 1372988_at | - | - |
| Rat Expression Set 230 | 1372988_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10920676 | - | - |
| Rat Genome U34 Set | rc_AI012014_at | - | - |
| Rat Genome U34 Set | rc_AI145642_at | - | - |
| Rat Genome U34 Set | rc_AI179930_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22319643 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366979 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49831 | - | - |

Cnot10
Genomic Map