Assay Details
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007802.3 | NP_001007803.1 | 3 - 4 | 353 | 59 | |
XM_006243969.2 | XP_006244031.1 | 2 - 3 | 375 | 59 | ||
GenBank mRNA | AY325262.1 | - | 3 - 4 | 353 | 59 | |
AY387088.1 | - | 2 - 3 | 313 | 59 | ||
BC107930.1 | - | 2 - 3 | 346 | 7130158 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
