Assay Details
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108182.1 | NP_001101652.1 | 12 - 13 | 1796 | 67 | |
XM_008766530.1 | XP_008764752.1 | 12 - 13 | 1541 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P134490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135569 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134490 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134490 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P134490 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P485556 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P187744 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56557 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21231 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41026 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15500 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18794 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10919814 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919810 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919813 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919808 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10919812 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10919804 | - | - |
Affymetrix Rat Clariom S Assay | PSR0800026306.rn.1 | TC0800003296.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800026308.rn.1 | TC0800003296.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800026307.rn.1 | TC0800003296.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10919804 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21121128 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373213 | - | - |
