Assay Details
| Gene Symbol: Amotl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031717.1 | NP_113905.1 | 5 - 6 | 1408 | 65 | |
| XM_006243680.2 | XP_006243742.1 | 5 - 6 | 1510 | 65 | ||
| GenBank mRNA | AF175969.1 | - | 4 - 5 | 545 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amotl2
Genomic Map