Assay Details
| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191883.1 | NP_001178812.1 | 11 - 12 | 976 | 57 | |
| XM_006243802.2 | XP_006243864.1 | 12 - 13 | 1472 | 57 | ||
| XM_006243803.2 | XP_006243865.1 | 12 - 13 | 1471 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcbp4
Genomic Map