Assay Details
| Gene Symbol: Zbtb38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766521.1 | XP_008764743.1 | 4 - 5 | 759 | 64 | |
| XM_008766522.1 | XP_008764744.1 | 5 - 6 | 770 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766521.1, XM_008766522.1 transcripts.
Set Membership: |
|

Zbtb38
Genomic Map