Assay Details
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109449.2 | NP_001102919.1 | 4 - 5 | 793 | 89 | |
XM_006243856.2 | XP_006243918.1 | 4 - 5 | 1068 | 89 | ||
XM_006243857.2 | XP_006243919.1 | 4 - 5 | 851 | 89 | ||
GenBank mRNA | BC105621.1 | - | 4 - 5 | 793 | 7454021 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
