Assay Details
| Gene Symbol: Il20rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173438.1 | NP_001166909.1 | 4 - 5 | 611 | 60 | |
| GenBank mRNA | DQ222846.1 | - | 4 - 5 | 611 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il20rb
Genomic Map