Assay Details
| Gene Symbol: Slc17a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009713.2 | NP_001009713.1 | 9 - 10 | 1303 | 74 | |
| GenBank mRNA | AY800277.1 | - | 9 - 10 | 1302 | 74 | |
| BC097482.1 | - | 9 - 10 | 1303 | 7459136 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc17a5
Genomic Map