Assay Details
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108194.1 | NP_001101664.1 | 1 - 2 | 365 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
response to oxidative stress
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
intracellular signal transduction
cellular hypotonic response
negative regulation of potassium ion transmembrane transporter activity
negative regulation of rubidium ion transmembrane transporter activity
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
protein phosphorylation
response to oxidative stress
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
intracellular signal transduction
cellular hypotonic response
negative regulation of potassium ion transmembrane transporter activity
negative regulation of rubidium ion transmembrane transporter activity
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P074599 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P852993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175175 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074599 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P852993 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074599 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P852993 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P852993 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074599 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P836863 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P852993 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74942 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13682 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2911 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2271 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95357 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10920859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920846 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920851 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920844 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920853 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920854 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920852 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920843 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920856 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920850 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920845 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920847 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920848 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920842 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920849 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920857 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920858 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10920841 | - | - |
Rat Genome 230 2.0 Array | 1372096_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372096_at | - | - |
Affymetrix HT Rat Focus Array | 1372096_at | - | - |
Affymetrix HT Rat Focus Array | 1372096_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800028825.rn.1 | TC0800003541.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800028826.rn.1 | TC0800003541.rn.1 | RTA TC |
Rat Expression Set 230 | 1372096_at | - | - |
Rat Expression Set 230 | 1372096_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10920841 | - | - |
Rat Genome U34 Set | rc_AA875244_at | - | - |
Rat Genome U34 Set | rc_AI059517_at | - | - |
Rat Genome U34 Set | rc_AI178912_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21970926 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370412 | - | - |
