Assay Details
| Gene Symbol: Acy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005383.1 | NP_001005383.1 | 4 - 5 | 353 | 66 | |
| XM_006243715.2 | XP_006243777.1 | 5 - 6 | 655 | 66 | ||
| XM_006243716.2 | XP_006243778.1 | 5 - 6 | 659 | 66 | ||
| GenBank mRNA | AY580164.1 | - | 4 - 5 | 353 | 66 | |
| BC078930.1 | - | 5 - 6 | 416 | 7107165 | 66 | |
| FQ232544.1 | - | 5 - 6 | 510 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acy1
Genomic Map