Assay Details
| Gene Symbol: Atp1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012913.1 | NP_037045.1 | 4 - 5 | 669 | 76 | |
| GenBank mRNA | BC061719.1 | - | 4 - 5 | 672 | 5598284 | 76 |
| D84450.1 | - | 4 - 5 | 669 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b3
Genomic Map