Assay Details
| Gene Symbol: Prss44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109298.1 | NP_001102768.1 | 1 - 2 | 344 | 84 | |
| XM_008766647.1 | XP_008764869.1 | 1 - 2 | 344 | 84 | ||
| GenBank mRNA | BC168977.1 | - | 1 - 2 | 323 | 7453880 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Prss44
Genomic Map