Assay Details
Gene Symbol: Prss44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109298.1 | NP_001102768.1 | 3 - 4 | 766 | 116 | |
XM_008766647.1 | XP_008764869.1 | 3 - 4 | 766 | 116 | ||
XM_008766648.1 | XP_008764870.1 | 3 - 4 | 442 | 116 | ||
GenBank mRNA | BC168977.1 | - | 3 - 4 | 745 | 7453880 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
