Assay Details
| Gene Symbol: Mrps22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077530.1 | NP_001070998.1 | 1 - 2 | 181 | 88 | |
| GenBank mRNA | BC105629.1 | - | 1 - 2 | 181 | 7322855 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps22
Genomic Map