Assay Details
Gene Symbol: Rasgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170531.1 | NP_001164002.1 | 3 - 4 | 512 | 54 | |
GenBank mRNA | CB607130.1 | - | 3 - 4 | 387 | 54 | |
CB714920.1 | - | 1 - 2 | 148 | 54 | ||
X67241.1 | - | 3 - 4 | 451 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
