Assay Details
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277283.1 | NP_001264212.1 | 1 - 2 | 355 | 102 | |
GenBank mRNA | BC159433.1 | - | 1 - 2 | 347 | 9093094 | 102 |
FQ215646.1 | - | 1 - 2 | 289 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
