Assay Details
| Gene Symbol: Clstn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134377.1 | NP_599204.1 | 4 - 5 | 647 | 64 | |
| XM_008766475.1 | XP_008764697.1 | 3 - 4 | 533 | 64 | ||
| GenBank mRNA | AJ427342.1 | - | 4 - 5 | 647 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clstn2
Genomic Map