Assay Details
| Gene Symbol: Bmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108168.1 | NP_001101638.1 | 3 - 4 | 854 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
type B pancreatic cell development
pattern specification process
negative regulation of cell proliferation
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of steroid biosynthetic process
BMP signaling pathway
male genitalia development
negative regulation of aldosterone biosynthetic process
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of insulin-like growth factor receptor signaling pathway
positive regulation of transcription from RNA polymerase II promoter
cell development
positive regulation of epithelial cell proliferation
SMAD protein signal transduction
positive regulation of dendrite development
negative regulation of cortisol biosynthetic process
positive regulation of DNA-dependent DNA replication
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
BMP receptor binding
skeletal system development
type B pancreatic cell development
pattern specification process
negative regulation of cell proliferation
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of steroid biosynthetic process
BMP signaling pathway
male genitalia development
negative regulation of aldosterone biosynthetic process
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of insulin-like growth factor receptor signaling pathway
positive regulation of transcription from RNA polymerase II promoter
cell development
positive regulation of epithelial cell proliferation
SMAD protein signal transduction
positive regulation of dendrite development
negative regulation of cortisol biosynthetic process
positive regulation of DNA-dependent DNA replication
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
BMP receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P335679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116383 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P335679 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P335679 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P335679 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P335679 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67133 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98679 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10911717 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911716 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911718 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10911711 | - | - |
| Rat Genome 230 2.0 Array | 1382657_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385127_at | - | - |
| Rat Genome 230 2.0 Array | 1385127_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382657_at | - | - |
| Affymetrix HT Rat Focus Array | 1382657_at | - | - |
| Affymetrix HT Rat Focus Array | 1382657_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800008315.rn.1 | TC0800001124.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382657_at | - | - |
| Rat Expression Set 230 | 1385127_at | - | - |
| Rat Expression Set 230 | 1382657_PM_at | - | - |
| Rat Expression Set 230 | 1385127_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10911711 | - | - |
| Rat Genome U34 Set | rc_AA946142_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21556012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373140 | - | - |

Bmp5
Genomic Map