Assay Details
Gene Symbol: Cmtm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198754.2 | NP_942049.1 | 1 - 2 | 497 | 83 | |
GenBank mRNA | AF538323.1 | - | 1 - 2 | 300 | 83 | |
BC088094.1 | - | 1 - 2 | 497 | 7325894 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
