Assay Details
| Gene Symbol: RGD1309079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134472.1 | NP_001127944.1 | 2 - 3 | 1170 | 116 | |
| GenBank mRNA | AY325189.1 | - | 293 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P029227 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P481892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110326 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P481892 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029227 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029227 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P481892 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P481892 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P029227 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P481892 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P144379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10919325 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919327 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919323 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10919313 | - | - |
| Rat Genome 230 2.0 Array | 1376223_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376223_at | - | - |
| Rat Genome 230 2.0 Array | 1381772_at | - | - |
| Rat Genome 230 2.0 Array | 1381772_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389199_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389199_at | - | - |
| Affymetrix HT Rat Focus Array | 1381772_at | - | - |
| Affymetrix HT Rat Focus Array | 1389199_at | - | - |
| Affymetrix HT Rat Focus Array | 1389199_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376223_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381772_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376223_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800025114.rn.1 | TC0800003142.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800025113.rn.1 | TC0800003142.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800025112.rn.1 | TC0800003142.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376223_PM_at | - | - |
| Rat Expression Set 230 | 1389199_PM_at | - | - |
| Rat Expression Set 230 | 1389199_at | - | - |
| Rat Expression Set 230 | 1376223_at | - | - |
| Rat Expression Set 230 | 1381772_PM_at | - | - |
| Rat Expression Set 230 | 1381772_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919313 | - | - |
| Rat Genome U34 Set | rc_AI073239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21564605 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361443 | - | - |

RGD1309079
Genomic Map