Assay Details
Gene Symbol: Elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126098.1 | NP_001119570.1 | 2 - 3 | 438 | 74 | |
XM_008765987.1 | XP_008764209.1 | 2 - 3 | 227 | 74 | ||
XM_008765988.1 | XP_008764210.1 | 2 - 3 | 264 | 74 | ||
XM_008765989.1 | XP_008764211.1 | 2 - 3 | 260 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
