Assay Details
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023987.3 | NP_076477.3 | 4 - 5 | 1184 | 65 | |
XM_006242499.2 | XP_006242561.1 | 3 - 4 | 3147 | 65 | ||
XM_006242500.2 | XP_006242562.1 | 4 - 5 | 1917 | 65 | ||
XM_006242502.2 | XP_006242564.1 | 3 - 4 | 1416 | 65 | ||
XM_006242503.2 | XP_006242565.1 | 3 - 4 | 1375 | 65 | ||
GenBank mRNA | AF183430.1 | - | 2 - 3 | 952 | 65 | |
BC083555.1 | - | 4 - 5 | 1184 | 7112819 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
