Assay Details
| Gene Symbol: Birc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021752.2 | NP_068520.2 | 4 - 5 | 1919 | 80 | |
| XM_006242494.2 | XP_006242556.1 | 5 - 6 | 2396 | 80 | ||
| XM_006242495.2 | XP_006242557.1 | 5 - 6 | 2597 | 80 | ||
| GenBank mRNA | AF183431.1 | - | 4 - 5 | 1277 | 80 | |
| AF190020.1 | - | 4 - 5 | 2291 | 80 | ||
| BC062055.1 | - | 4 - 5 | 1919 | 5598786 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Birc2
Genomic Map