Assay Details
Gene Symbol: Kank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270413.1 | NP_001257342.1 | 8 - 9 | 2192 | 60 | |
XM_006242589.2 | XP_006242651.1 | 9 - 10 | 2597 | 60 | ||
XM_006242590.2 | XP_006242652.1 | 8 - 9 | 2452 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
