Assay Details
| Gene Symbol: RGD1309534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014206.1 | NP_001014228.1 | 2 - 3 | 171 | 81 | |
| GenBank mRNA | BC085915.1 | - | 2 - 3 | 171 | 7107444 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1309534
Genomic Map