Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198050.1 | NP_932167.1 | 16 - 17 | 3700 | 101 | |
| XM_006253556.2 | XP_006253618.1 | 16 - 17 | 3844 | 101 | ||
| XM_006253557.2 | XP_006253619.1 | 15 - 16 | 3700 | 101 | ||
| XM_008771424.1 | XP_008769646.1 | 16 - 17 | 3868 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P280712 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P16066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140796 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16066 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P280712 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P280712 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16066 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P280712 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P16066 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16066 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P280712 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3687 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90839 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4200 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52758 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10793859 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793847 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793853 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793863 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793860 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793856 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793864 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793850 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793865 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793857 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793855 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793861 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793848 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793852 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793862 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793851 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793849 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793854 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793858 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10793846 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700016248.rn.1 | TC1700002383.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700016249.rn.1 | TC1700002383.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700016247.rn.1 | TC1700002383.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10793846 | - | - |
| Rat Genome U34 Set | AF035954_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21362125 | - | - |

Kif27
Genomic Map