Assay Details
| Gene Symbol: Ctsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017121.1 | NP_058817.1 | 4 - 5 | 476 | 71 | |
| XM_008771439.1 | XP_008769661.1 | 3 - 4 | 391 | 71 | ||
| GenBank mRNA | AF310623.2 | - | 4 - 5 | 476 | 71 | |
| BC097263.1 | - | 4 - 5 | 459 | 7460052 | 71 | |
| L14776.1 | - | 1 - 2 | 108 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsj
Genomic Map