Assay Details
| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106630.1 | NP_001100100.1 | 3 - 4 | 1450 | 125 | |
| XM_006237719.2 | XP_006237781.1 | 3 - 4 | 1450 | 125 | ||
| XM_006237720.2 | XP_006237782.1 | 3 - 4 | 1450 | 125 | ||
| XM_006237721.2 | XP_006237783.1 | 3 - 4 | 1450 | 125 | ||
| XM_006237722.2 | XP_006237784.1 | 3 - 4 | 1450 | 125 | ||
| XM_006237723.2 | XP_006237785.1 | 3 - 4 | 1450 | 125 | ||
| XM_008763475.1 | XP_008761697.1 | 3 - 4 | 1450 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jph1
Genomic Map