Assay Details
| Gene Symbol: Clvs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108969.1 | NP_001102439.1 | 6 - 7 | 1468 | 89 | |
| GenBank mRNA | FQ214563.1 | - | 5 - 6 | 1293 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P290953 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P111212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69554 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P290953 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P111212 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P290953 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P111212 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P111212 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P290953 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P111212 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P188702 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P290953 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22130 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101222 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63511 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35629 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10867592 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867588 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867590 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867591 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867589 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10867587 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500001178.rn.1 | TC0500000177.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10867587 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21902116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365631 | - | - |

Clvs1
Genomic Map