Assay Details
Gene Symbol: Cpeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770226.1 | XP_008768448.1 | 1719 | 87 | ||
GenBank mRNA | JF973322.1 | - | 1629 | 87 | ||
JF973323.1 | - | 1719 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
