Assay Details
Gene Symbol: Emid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109467.1 | NP_001102937.1 | 4 - 5 | 664 | 63 | |
XM_008770367.1 | XP_008768589.1 | 4 - 5 | 417 | 63 | ||
XM_008770368.1 | XP_008768590.1 | 4 - 5 | 417 | 63 | ||
XM_008770369.1 | XP_008768591.1 | 4 - 5 | 417 | 63 | ||
XM_008770370.1 | XP_008768592.1 | 4 - 5 | 417 | 63 | ||
XM_008770371.1 | XP_008768593.1 | 4 - 5 | 435 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
