Assay Details
Gene Symbol: Kcnj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031610.3 | NP_113798.1 | 2 - 3 | 974 | 61 | |
GenBank mRNA | L25264.1 | - | 2 - 3 | 974 | 61 | |
U01071.1 | - | 2 - 3 | 947 | 61 | ||
U01141.1 | - | 2 - 3 | 988 | 61 | ||
U09243.1 | - | 1102 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P452282 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P452163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155318 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P452163 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P452282 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P452282 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P452163 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P452282 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P452163 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P452282 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12612 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P452163 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247906 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6921 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91781 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71542 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45538 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98015 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 347 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28707 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 294 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1288 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97017 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24588 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71160 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87709 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10836272 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836275 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836273 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836274 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10836271 | - | - |
Rat Genome 230 2.0 Array | 1369740_at | - | - |
Rat Genome 230 2.0 Array | 1369742_at | - | - |
Rat Genome 230 2.0 Array | 1369741_at | - | - |
Rat Genome 230 2.0 Array | 1369742_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369740_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369741_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388292_at | - | - |
Rat Genome 230 2.0 Array | 1388292_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369742_at | - | - |
Affymetrix HT Rat Focus Array | 1369742_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369741_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369740_at | - | - |
Affymetrix HT Rat Focus Array | 1369741_at | - | - |
Affymetrix HT Rat Focus Array | 1388292_at | - | - |
Affymetrix HT Rat Focus Array | 1369740_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388292_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300004736.rn.1 | TC0300000610.rn.1 | RTA TC |
Rat Expression Set 230 | 1369742_PM_at | - | - |
Rat Expression Set 230 | 1369740_PM_at | - | - |
Rat Expression Set 230 | 1369741_PM_at | - | - |
Rat Expression Set 230 | 1388292_at | - | - |
Rat Expression Set 230 | 1388292_PM_at | - | - |
Rat Expression Set 230 | 1369742_at | - | - |
Rat Expression Set 230 | 1369740_at | - | - |
Rat Expression Set 230 | 1369741_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836271 | - | - |
Rat Genome U34 Set | U09243_at | - | - |
Rat Genome U34 Set | U72410_s_at | - | - |
Rat Genome U34 Set | U72410_s_at | - | - |
Rat Genome U34 Set | U09243_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373503 | - | - |
RatRef-12 Expression BeadChip | ILMN_64776 | - | - |
