Assay Details
Gene Symbol: Galnt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199106.1 | NP_954537.1 | 4 - 5 | 779 | 61 | |
XM_008761746.1 | XP_008759968.1 | 2 - 3 | 378 | 61 | ||
GenBank mRNA | AY371923.1 | - | 4 - 5 | 779 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
