Assay Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107842.1 | NP_001101312.1 | 13 - 14 | 1447 | 65 | |
| XM_006234096.2 | XP_006234158.1 | 13 - 14 | 1501 | 65 | ||
| XM_006234097.2 | XP_006234159.1 | 13 - 14 | 1457 | 65 | ||
| XM_006234099.2 | XP_006234161.1 | 13 - 14 | 1501 | 65 | ||
| XM_008761758.1 | XP_008759980.1 | 13 - 14 | 1503 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Golga1
Genomic Map